2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-(Oxiran-2-ylmethyl)isoindoline-1,3-dione, N-(2,3-Epoxypropyl)phthalimide
170040100
170040250
ACRO170040100EA
117
EUR
ACRO170040100
ACRO170040250
2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione 95%
2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione
Formel:
C₁₁H₉NO₃ Schmelzpunkt: 94…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005896 CAS-Nummer: 5455-98-1 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 92 to 102 °C |
GC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...