1-(4-Methoxyphenyl)cyclopropanecarbonitrile 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-(4-Methoxyphenyl)-1-cyclopropanecarbonitrile

170620250
ACRO170620250EA 103 EUR
ACRO170620250
1-(4-Methoxyphenyl)cyclopropanecarbonitrile 96%
1-(4-Methoxyphenyl)cyclopropanecarbonitrile

Appearance: Clear colorless to yellow Liquid

Formel: C₁₁H₁₁NO
Molecular Weight: 173,215 g/mol
Dichte: 1,08 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00019212
CAS-Nummer: 16728-00-0
UN: 3276
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5415 to 1.5435 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR