1-(4-Methylphenyl)cyclopropancarbonsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(p-Tolyl)cyclopropanecarboxylic acid

170700100
ACRO170700100EA 42.2 EUR
ACRO170700100
1-(4-Methylphenyl)cyclopropancarbonsäure 98%
1-(4-Methylphenyl)cyclopropanecarboxylic acid
Formel: C₁₁H₁₂O₂
Molecular Weight: 176.2100 g/mol
Schmelzpunkt: 109…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066918
CAS-Nummer: 83846-66-6
EINECS: 281-043-4

Bestellen

Spezifikation Testergebnisse

Appearance Beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 109 to 114 °C
Titration with NaOH ≥97.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR