ɑ-Bromocinnamaldehyde 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Bromo-3-phenyl-2-propenal, 2-Bromo-3-phenylacrylaldehyde, Alpha-Bromocinnamaldehyde

171272500
ACRO171272500EA 65 EUR
ACRO171272500
ɑ-Bromocinnamaldehyde 98%
alpha-Bromocinnamaldehyde

CAS No.: 5443-49-2

Formel: C₉H₇BrO
Molecular Weight: 211,06 g/mol
Schmelzpunkt: 68…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006965
CAS-Nummer: 5443-49-2

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to ochre Crystalline powder
Infrared spectrum Conforms
Melting point 68 to 72 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR