1,3-Diethyl-2-thiobarbitursäure 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
171740500
171740050
171742500
ACRO171740500EA
128
EUR
ACRO171740500
ACRO171740050
ACRO171742500
1,3-Diethyl-2-thiobarbitursäure 99%
1,3-Diethyl-2-thiobarbitursäure
Formel:
C₈H₁₂N₂O₂S Schmelzpunkt: 106…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006676 CAS-Nummer: 5217-47-0 EINECS: 226-010-7 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 105 to 109 °C |
GC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...