DL-Tryptophan 98%

Lieferant: Thermo Fisher Scientific

Synonyme: H-DL-Trp-OH, (+/-)-2-Amino-3-(3-indolyl)propionic acid, 2-Amino-3-(1H-indol-3-yl)propanoic acid

172110250 172111000
ACRO172110250EA 49.9 EUR
ACRO172110250 ACRO172111000
DL-Tryptophan 98%
DL-Tryptophan
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204.23 g/mol
Schmelzpunkt: 289…290 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00064339
CAS-Nummer: 54-12-6
EINECS: 200-194-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.8 % (105°C, 3 h) (Vacuum)
Solubility (1 % in 1 M HCl) Clear to slightly hazy colorles to yellow
Titration with HClO4 ≥97.5 %
Other amino acids (TLC) Not detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR