2-Picolylchlorid Hydrochlorid 98%

Lieferant: Thermo Fisher Scientific
Gefahr

172120050 172120250 172121000
ACRO172120050EA 39.6 EUR
ACRO172120050 ACRO172120250 ACRO172121000
2-Picolylchlorid Hydrochlorid 98%
2-Picolylchlorid Hydrochlorid
Formel: C₆H₇Cl₂N
Schmelzpunkt: 119…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012811
CAS-Nummer: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow-green to light brown Crystalline mass
Infrared spectrum Conforms
Melting point 119 to 125 °C
Titration with HClO4 ≥97.5 %
Specific optical rotation -115° to -130° (20 °C, 589 nm) (c=0.8 in EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR