2-Picolylchlorid Hydrochlorid 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
172120050
172120250
172121000
ACRO172120050EA
39.6
EUR
ACRO172120050
ACRO172120250
ACRO172121000
2-Picolylchlorid Hydrochlorid 98%
2-Picolylchlorid Hydrochlorid
Formel:
C₆H₇Cl₂N Schmelzpunkt: 119…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012811 CAS-Nummer: 6959-47-3 EINECS: 230-149-9 UN: 3261 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Yellow-green to light brown Crystalline mass |
Infrared spectrum | Conforms |
Melting point | 119 to 125 °C |
Titration with HClO4 | ≥97.5 % |
Specific optical rotation | -115° to -130° (20 °C, 589 nm) (c=0.8 in EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...