4-Chlor-7-nitrobenzofurazan 98%

Lieferant: Thermo Fisher Scientific
Achtung

172390010 172390050 172390250
ACRO172390010EA 29.3 EUR
ACRO172390010 ACRO172390050 ACRO172390250
4-Chlor-7-nitrobenzofurazan 98%
4-Chlor-7-nitrobenzofurazan
Formel: C₆H₂ClN₃O₃
Schmelzpunkt: 96…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005808
CAS-Nummer: 10199-89-0
EINECS: 233-496-4

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 99 °C
Sulfated ash ≤0.1 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR