2-Aminobenzimidazol 99+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Benzimidazol-2-ylamin, 2-Amino-1H-benzimidazol

174020250 174021000
ACRO174020250EA 85.2 EUR
ACRO174020250 ACRO174021000
2-Aminobenzimidazol 99+%
2-Aminobenzimidazole
Formel: C₇H₇N₃
Molecular Weight: 133.15 g/mol
Siedepunkt: 381
Schmelzpunkt: 228…232 °C
MDL: MFCD00005596
CAS-Nummer: 934-32-7
EINECS: 213-280-6

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to beige to light brown Crystalline powder or crystals or flakes (May darken on storage)
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR