L(+)-Rhamnose Monohydrat 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Isodulcit, 6-Desoxy-L(+)-mannose Monohydrat

174080250 174081000 174085000
ACRO174080250EA 187 EUR
ACRO174080250 ACRO174081000 ACRO174085000
L(+)-Rhamnose Monohydrat 99%
L(+)-Rhamnose Monohydrat
Formel: C₆H₁₂O₅·1H₂O
Molecular Weight: 182.17 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149363
CAS-Nummer: 10030-85-0
EINECS: 222-793-4
Merck Index: 12,08338

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Water 9.0 to 11.0 % (K.F.)
Heavy metals ≤20 ppm
HPLC ≥98.5 % (ELSD)
Residue after ignition ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR