Quecksilber(II)acetat 98%

Lieferant: Thermo Fisher Scientific
Gefahr

175021000 175025000 175020025 175020050
ACRO175021000EA 70.6 EUR
ACRO175021000 ACRO175025000 ACRO175020025 ACRO175020050
Quecksilber(II)acetat 98%
Quecksilber(II)acetat
Formel: (H₃CCOO)₂Hg
Schmelzpunkt: 179…182 °C
Dichte: 3,27 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012165
CAS-Nummer: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 182 °C
Titration Complexometric ≥97.5 %
Chloride (Cl) ≤300 ppm
Sulfate (SO4) ≤100 ppm
Iron (Fe) ≤50 ppm
Mercurous Mercury (as Hg) ≤8000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR