3-Chinolincarbaldehyd 97+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Chinolincarboxaldehyd
175690010
175690050
ACRO175690010EA
98.8
EUR
ACRO175690010
ACRO175690050
3-Chinolincarbaldehyd 97+%
3-Chinolincarbaldehyd
Formel:
C₁₀H₇NO Molecular Weight: 157.17 g/mol Schmelzpunkt: 66…69 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006768 CAS-Nummer: 13669-42-6 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 66 to 69 °C |
GC | ≥97.0 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...