Tetrabutylammoniumhydroxid 40% (w/w) 1.5 M in Wasser
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
176610010
176610500
176612500
176610025
ACRO176610010EA
409
EUR
ACRO176610010
ACRO176610500
ACRO176612500
ACRO176610025
Tetrabutylammoniumhydroxid 40% (w/w) 1.5 M in Wasser
Tetrabutylammoniumhydroxid
Formel:
C₁₆H₃₇NO Dichte: 0,995 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009425 CAS-Nummer: 2052-49-5 EINECS: 218-147-6 UN: 3267 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear solution |
Color scale | ≤150 APHA |
Titration with HCl | 38 to 42 % |
Solubility | (5% in water) Clear colorless |
Specific optical rotation | +20.0° to +24.0° (20°C, 589 nm) (c=1%, water) |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...