1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
177480050
177481000
177485000
ACRO177480050EA
36.7
EUR
ACRO177480050
ACRO177481000
ACRO177485000
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) 99%
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture)
Appearance: Clear colorless Liquid
Formel:
C₈H₁₈N₂ Siedepunkt: 220 °C (760 mmHg) Schmelzpunkt: –70 °C Dichte: 0,94 g/cm³ Flammpunkt: 106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001522 CAS-Nummer: 2579-20-6 UN: 3267 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4920 to 1.4940 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...