3,3-Dimethylacryloyl chloride 97%

Lieferant: Thermo Fisher Scientific
Gefahr

179730050 179730250
ACRO179730050EA 32.9 EUR
ACRO179730050 ACRO179730250
3,3-Dimethylacryloyl chloride 97%
3,3-Dimethylacryloyl chloride

Appearance: Clear colorless to yellow Liquid

Formel: C₅H₇ClO
Siedepunkt: 145…147 °C (760 mmHg)
Dichte: 1,06 g/cm³
Flammpunkt: 51 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00000730
CAS-Nummer: 3350-78-5
UN: 2920
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4780 (20°C, 589 nm)
GC ≥96.0 %
Titration with NaOH 98.0 to 100.5 % (dried substance)
Chloride (Cl) ≤0.04 %
Sulfate (SO4) ≤0.1 %
Arsenic (As) ≤4 ppm
Iron (Fe) ≤40 ppm
Heavy metals (as Pb) ≤20 ppm
Impurity ≤2.5 % (Monosodium phosphate)
Insoluble matter ≤0.4 %
Reducing substances Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR