DL-Valinol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-3-methylbutan-1-ol, DL-2-Amino-3-methyl-1-butanol, 2-Amino-3-methyl-1-butanol

180420050
ACRO180420050EA 74.4 EUR
ACRO180420050
DL-Valinol 97%
DL-Valinol

Appearance: Clear yellow Liquid

Formel: C₅H₁₃NO
Molecular Weight: 103.17 g/mol
Siedepunkt: 75…77 °C (8 mmHg)
Dichte: 0.93 g/cm³
Flammpunkt: 90 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00004730
CAS-Nummer: 16369-05-4

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4570 (20°C, 589 nm)
GC ≥96.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR