Pyren 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
180830250
180831000
180835000
ACRO180830250EA
35
EUR
ACRO180830250
ACRO180831000
ACRO180835000
Pyren 98%
Pyrene
Formel:
C₁₆H₁₀ Molecular Weight: 202.26 g/mol Siedepunkt: 393 °C (760 mmHg) Schmelzpunkt: 148…152 °C Flammpunkt: 210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004136 CAS-Nummer: 129-00-0 EINECS: 204-927-3 UN: 3077 ADR: 9 III Merck Index: 12,08147 |
|
Spezifikation Testergebnisse
Appearance | Light yellow to yellow to yellow-green Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 147 to 152 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...