(S)-(-)-Nicotin 99+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
L(-)-Nicotin, (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridin, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridin
181420050
181420250
181421000
181425000
ACRO181420050EA
33.6
EUR
ACRO181420050
ACRO181420250
ACRO181421000
ACRO181425000
(S)-(-)-Nicotin 99+%
(S)-(-)-Nicotin
Formel:
C₁₀H₁₄N₂ Siedepunkt: 247 °C (760 mmHg) Schmelzpunkt: –80 °C Dichte: 1,015 g/cm³ Flammpunkt: 101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006369 CAS-Nummer: 54-11-5 EINECS: 200-193-3 UN: 1654 ADR: 6.1,II Merck Index: 12,06611 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5255 to 1.5285 (20°C, 589 nm) |
Water | ≤0.5 % (K.F.) |
GC | ≥99.0 % |
Specific optical rotation | -140° to -152° (20°C, 589 nm) (c=2,absolute ethanol) (on anhydrous basis) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...