N,N-Dimethyl-1,4-phenylendiammoniumsulfat 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
181430250
181431000
ACRO181430250EA
72.2
EUR
ACRO181430250
ACRO181431000
N,N-Dimethyl-1,4-phenylendiammoniumsulfat 99%
N,N-Dimethyl-1,4-phenylendiammoniumsulfat
Formel:
C₈H₁₄N₂O₄S Schmelzpunkt: 234…236 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00012992 CAS-Nummer: 536-47-0 EINECS: 208-636-2 UN: 2811 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | White to grey to brown Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Titration with NaOH | ≥98.5 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...