Trimethylene sulphide

Lieferant: Thermo Fisher Scientific
Gefahr

182910050 182910250
ACRO182910050EA 78.8 EUR
ACRO182910050 ACRO182910250
Trimethylene sulphide
Trimethylene sulphide

  • Building Block for chemical synthesis

Formel: C₃H₆S
Siedepunkt: 94…94,5 °C (760 mmHg)
Dichte: 1,028
Flammpunkt: –2 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042751
CAS-Nummer: 287-27-4
UN: 1993
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5040 to 1.5140 (20°C, 589 nm)
GC ≥96.0 %
UV at 290 nm A: ≤0.1 (33% W/W)
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR