Poly(caprolactone)diol, M.W. ∼ 2000

Lieferant: Thermo Fisher Scientific

183212500 183215000 183210025
ACRO183212500EA 167 EUR
ACRO183212500 ACRO183215000 ACRO183210025
Poly(caprolactone)diol, M.W. ∼ 2000
Poly(caprolactone)diol

CAS No.: 36890-68-3

Dichte: 1,07 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD23713083
CAS-Nummer: 36890-68-3

Bestellen

Spezifikation Testergebnisse

Appearance White Waxy solid
Infrared spectrum Conforms
Water ≤2 % (K.F.)
Molecular number 1500 to 2500


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR