Polyvinylacetat, M.W. ∼100000
Lieferant: Thermo Fisher Scientific
183265000
ACRO183265000EA
125
EUR
ACRO183265000
Polyvinylacetat, M.W. ∼100000
Polyvinylacetat
Formel:
[CH₂CH(O₂CCH₃)]n Dichte: 1,18 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084457 CAS-Nummer: 9003-20-7 |
Spezifikation Testergebnisse
Appearance | Clear Beads |
Infrared spectrum | Conforms |
Loss on drying | ≤5 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...