(±)-4-Phenyl-2-butanol 97%
Lieferant: Thermo Fisher Scientific
183400250
183400050
ACRO183400250EA
96.5
EUR
ACRO183400250
ACRO183400050
(±)-4-Phenyl-2-butanol 97%
(±)-4-Phenyl-2-butanol
Formel:
(C₆H₅)CH₂CH₂CH(OH)CH₃ Molecular Weight: 150.22 g/mol Siedepunkt: 132 °C (14 mmHg) Dichte: 0.98 g/cm³ Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00044349 CAS-Nummer: 2344-70-9 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5150 to 1.5180 (20°C, 589 nm) |
GC | ≥96.0 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...