Plumbagin 99+%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Hydroxy-2-methylnaphthalene-1,4-dione, 5-Hydroxy-2-methyl-1,4-naphthoquinone

184040010
ACRO184040010EA 184 EUR
ACRO184040010
Plumbagin 99+%
Plumbagin

Appearance: Orange Crystalline powder or crystals

Formel: C₁₁H₈O₃
Molecular Weight: 188,18 g/mol
Schmelzpunkt: 75…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001682
CAS-Nummer: 481-42-5
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Orange Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 75 to 78 °C
GC ≥99.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR