O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 98%

Lieferant: Thermo Fisher Scientific
Achtung

186710010 186710050 186712500
ACRO186710010EA 139 EUR
ACRO186710010 ACRO186710050 ACRO186712500
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 98%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride
Formel: C₇H₄F₅NO
Lagertemperatur: Kühlschrank
MDL: MFCD00012953
CAS-Nummer: 57981-02-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder, crystals, flakes and/or chunks
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Composition ≤2 % (CoCl2) Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR