N-(tert-Butyl)hydroxylamine hydrochloride 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-(tert-Butyl) hydroxylamine hydrochloride
186910010
ACRO186910010EA
46.6
EUR
ACRO186910010
N-(tert-Butyl)hydroxylamine hydrochloride 98%
N-(tert-Butyl)hydroxylamine hydrochloride
Formel:
C₄H₁₁NO Molecular Weight: 125,60 g/mol Schmelzpunkt: 176…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012598 CAS-Nummer: 57497-39-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 175 to 185 °C |
Titration Argentometric | ≥97.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...