tert-Butylhydrazin Hydrochlorid 98%
Lieferant: Thermo Scientific
|
Achtung
|
187081000
ACRO187081000EA
91.3
EUR
ACRO187081000
tert-Butylhydrazin Hydrochlorid 98%
tert-Butylhydrazin Hydrochlorid
Formel:
C₄H₁₃ClN₂ Molecular Weight: 124,61 g/mol Schmelzpunkt: 190…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012947 CAS-Nummer: 7400-27-3 EINECS: 231-001-6 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 190 to 194 °C |
Titration with HClO4 | ≥97.5 % |
Additional info | Denatured with 1% iPr-OH, 1% MEK and 0.001% Bitrex |
E/E | 99.8 to 100.0 % v/v (calculated) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...