2,5-Dihydroxy-p-benzochinon 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,5-Dihydroxy-1,4-benzoquinone
187460250
187461000
ACRO187460250EA
66.8
EUR
ACRO187460250
ACRO187461000
2,5-Dihydroxy-p-benzochinon 98%
2,5-Dihydroxy-p-benzochinon
Formel:
C₆H₄O₄ Molecular Weight: 140.1 g/mol Schmelzpunkt: 220 °C (Subl.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001598 CAS-Nummer: 615-94-1 EINECS: 210-454-3 UN: 2811 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | Ochre to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Titration with NaOH | 9.3 to 9.7 % (fumaric acid dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...