2,5-Dihydroxy-p-benzochinon 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,5-Dihydroxy-1,4-benzoquinone

187460250 187461000
ACRO187460250EA 66.8 EUR
ACRO187460250 ACRO187461000
2,5-Dihydroxy-p-benzochinon 98%
2,5-Dihydroxy-p-benzochinon
Formel: C₆H₄O₄
Molecular Weight: 140.1 g/mol
Schmelzpunkt: 220 °C (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001598
CAS-Nummer: 615-94-1
EINECS: 210-454-3
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Ochre to brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH 9.3 to 9.7 % (fumaric acid dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR