4-Morpholinoanilin 98+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(4-Morpholinyl)aniline

188650050 188650250
ACRO188650050EA 43.1 EUR
ACRO188650050 ACRO188650250
4-Morpholinoanilin 98+%
4-Morpholinoanilin
Formel: C₁₀H₁₄N₂O
Molecular Weight: 178.23 g/mol
Schmelzpunkt: 130…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006169
CAS-Nummer: 2524-67-6
EINECS: 219-760-1
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Brown to purple Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 132 °C
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR