Homoveratrylalkohol 98%
Lieferant: Thermo Scientific
Synonyme:
Homoveratryl Alcohol, 3,4-Dimethoxyphenethyl alcohol, 3,4-Dimethoxybenzeneethanol, 3,4-Dimethoxyphenethyl Alcohol
189000100
ACRO189000100EA
57.6
EUR
ACRO189000100
Homoveratrylalkohol 98%
2-(3,4-Dimethoxyphenyl)ethanol
CAS No.: 7417-21-2
Formel:
C₁₀H₁₄O₃ Molecular Weight: 182,22 g/mol Siedepunkt: 172…174 °C (17 mmHg) Schmelzpunkt: 44…48 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002894 CAS-Nummer: 7417-21-2 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 44 to 48 °C |
GC | ≥97.5 % |
UV | at 270 nm A: ≤0.1 |
Titration with NaOH | ≤99.75 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...