ω-Brom-2,4-dimethoxyacetophenon 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,4-Dimethoxyphenacylbromid, 2-Brom-2',4'-dimethoxyacetophenon, α-Brom-2,4-dimethoxyacetophenon, 2-Brom-1-(2,4-dimethoxyphenyl)ethan-1-on
190010010
ACRO190010010EA
76.3
EUR
ACRO190010010
ω-Brom-2,4-dimethoxyacetophenon 98%
ω-Brom-2,4-dimethoxyacetophenon
Formel:
C₁₀H₁₁BrO₃ Molecular Weight: 259,1 g/mol Siedepunkt: 315 °C (1013 hPa) Schmelzpunkt: 101…105 °C |
MDL:
MFCD00000197 CAS-Nummer: 60965-26-6 EINECS: 262-542-6 UN: 3261 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 101 to 105 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...