1-Bromoctadecan 96%

Lieferant: Thermo Fisher Scientific

Synonyme: n-Octadecylbromid

190021000 190025000 190020025
ACRO190021000EA 30.4 EUR
ACRO190021000 ACRO190025000 ACRO190020025
1-Bromoctadecan 96%
1-Bromoctadecan
Formel: BrH₂C(CH₂)₁₆CH₃
Molecular Weight: 333.4 g/mol
MDL: MFCD00000231
CAS-Nummer: 112-89-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Fused solid or liquid
Infrared spectrum Conforms
Melting point 25 to 31 °C
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR