Sudan II, rein
Lieferant: Thermo Scientific
Synonyme: Sudan® II (Orange), 1-(2,4-Dimethylphenylazo)-2-naphthol, Solvent Orange 7, Sudan Orange
190150250
ACRO190150250EA 38.3 EUR
ACRO190150250
Sudan II, rein
Sudan II
Bestellen
Spezifikation Testergebnisse
Appearance Red to orange-brown Powder
Infrared spectrum Conforms
UV Lambda max at approximately 420 and 493 nm (in methanol)
Specific extinction E (1%/1cm) ≥500 (at 493 nm, in methanol)