Sudanorange G, pure

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(Phenylazo)resorcin, Solvent Orange 1

190170050 190170250 190170010 190171000
ACRO190170050EA 23 EUR
ACRO190170050 ACRO190170250 ACRO190170010 ACRO190171000
Sudanorange G, pure
Sudan Orange G
Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214.22 g/mol
Siedepunkt: 371
Schmelzpunkt: 143…146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002275
CAS-Nummer: 2051-85-6
EINECS: 218-131-9

Bestellen

Spezifikation Testergebnisse

Appearance Orange-red Powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR