Tropaeolin O, pure

Lieferant: Thermo Fisher Scientific

Synonyme: Sodium 2,4-Dihydroxyazobenzene-4'-sulfonate, 2,4-Dihydroxyazobenzene-4'-sulfonic Acid Sodium Salt, Acid Orange 6, Resorcinol Yellow

190180250 190181000
ACRO190180250EA 65.4 EUR
ACRO190180250 ACRO190181000
Tropaeolin O, pure
Tropaeolin O

CAS No.: 547-57-9

Formel: C₁₂H₉N₂O₅SNa
Molecular Weight: 316.26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007499
CAS-Nummer: 547-57-9

Bestellen

Spezifikation Testergebnisse

Appearance Rust brown to dark brown Powder
Infrared spectrum Conforms
Solubility (in water) Passes test
Visual transition interval from pH 11.1 (yellow) to pH 12.7 (orange)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR