Ruthenium red tetrahydrate, rein

Lieferant: Thermo Fisher Scientific
Achtung

191730010 191730050
ACRO191730010EA 105 EUR
ACRO191730010 ACRO191730050
Ruthenium red tetrahydrate, rein
Ruthenium red tetrahydrate

CAS No.: 12790-48-6

Formel: Cl₆H₄₂N₁₄O₂Ru₃·4H₂O
Lagertemperatur: Kühlschrank
MDL: MFCD00167176
CAS-Nummer: 12790-48-6

Bestellen

Spezifikation Testergebnisse

Appearance Dark red-purple Crystalline powder
Assay 34.6 to 36.1 % (Ru)
UV Lambda max 530 to 536 nm
Molar extinction coefficient ≥8000 l/(mole.cm) (lambda max., 0.02 g/l H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR