Cäsiumcarbonat 99.5% zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

192040250 192041000 192040050 192045000 192040025
ACRO192040250EA 31.8 EUR
ACRO192040250 ACRO192041000 ACRO192040050 ACRO192045000 ACRO192040025
Cäsiumcarbonat 99.5% zur Analyse
Cesium carbonate
Formel: Cs₂CO₃
Molecular Weight: 325.82 g/mol
Schmelzpunkt: >400 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010957
CAS-Nummer: 534-17-8
EINECS: 208-591-9
Merck Index: 13,02020

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Assay ≥99.4 % (By difference)
Loss on drying ≤0.5 % (500°C)
Chloride (Cl) ≤300 ppm
Sulfate (SO4) ≤500 ppm
Aluminium (Al) ≤100 ppm
Barium (Ba) ≤100 ppm
Calcium (Ca) ≤20 ppm
Potassium (K) ≤400 ppm
Rubidium (Rb) ≤3000 ppm
Sodium (Na) ≤300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR