Cobalt(II)carbonat Hydrat

Lieferant: Thermo Fisher Scientific
Gefahr

192100010 192100500 192102500
ACRO192100010EA 573 EUR
ACRO192100010 ACRO192100500 ACRO192102500
Cobalt(II)carbonat Hydrat
Cobalt(II)carbonat Hydrat
Formel: CCoO₃·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00149654
CAS-Nummer: 57454-67-8
EINECS: 208-169-4
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance Pink to red to violet Powder
Titration Complexometric ≥45.5 % (Cobalt)
Cadmium (Cd) ≤50 ppm
Copper (Cu) ≤100 ppm
Iron (Fe) ≤100 ppm
Insoluble matter ≤0.05 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR