Ammoniumiodid ACS

Lieferant: Thermo Fisher Scientific
Achtung

193210010 193210500 193212500
ACRO193210010EA 453 EUR
ACRO193210010 ACRO193210500 ACRO193212500
Ammoniumiodid ACS
Ammoniumiodid
Formel: NH₄I
Schmelzpunkt: 551 °C (Subl.)
Dichte: 2,51 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011424
CAS-Nummer: 12027-06-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or crystals (May turn yellow on storage)
Residue after ignition ≤0.05 %
Titration ≥99.0 %
Chloride and Bromide ≤0.005 % (as Cl)
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.05 %
Barium (Ba) ≤0.002 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR