Praseodymtrinitrat Pentahydrat 99.9% Trace metals basis
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Praseodymium (III) nitrate pentahydrate
194590500
ACRO194590500EA
69.8
EUR
ACRO194590500
Praseodymtrinitrat Pentahydrat 99.9% Trace metals basis
Praseodymtrinitrat Pentahydrat
Formel:
N₃O₉Pr·5H₂O Molecular Weight: 417 Schmelzpunkt: 69 Dichte: 2.233 |
CAS-Nummer:
14483-17-1 EINECS: 233-796-5 UN: 1477 ADR: 5.1 II |
Spezifikation Testergebnisse
Appearance | Green Crystalline powder |
Total conc. of imp.as REO | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...