Dimethyl-2-aminoterephthalat 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate
194780250
194780500
ACRO194780250EA
51.3
EUR
ACRO194780250
ACRO194780500
Dimethyl-2-aminoterephthalat 99%
Dimethyl-2-aminoterephthalat
Formel:
C₁₀H₁₁NO₄ Schmelzpunkt: 132…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008427 CAS-Nummer: 5372-81-6 EINECS: 226-364-2 |
Spezifikation Testergebnisse
Appearance | Yellow to beige to light green to brown Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 131 to 134 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...