Dimethyl-2-aminoterephthalat 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate

194780250 194780500
ACRO194780250EA 51.3 EUR
ACRO194780250 ACRO194780500
Dimethyl-2-aminoterephthalat 99%
Dimethyl-2-aminoterephthalat
Formel: C₁₀H₁₁NO₄
Schmelzpunkt: 132…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008427
CAS-Nummer: 5372-81-6
EINECS: 226-364-2

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to beige to light green to brown Powder or crystals
Infrared spectrum Conforms
Melting point 131 to 134 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR