Platin(II)chlorid (≥73% Pt)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
195380010
195380050
195380250
195382500
ACRO195380010EA
170
EUR
ACRO195380010
ACRO195380050
ACRO195380250
ACRO195382500
Platin(II)chlorid (≥73% Pt)
Platin(II)chlorid
Formel:
Cl₂Pt Dichte: 6,05 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011181 CAS-Nummer: 10025-65-7 EINECS: 233-034-1 UN: 3260 ADR: 8,III Merck Index: 12,07687 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...