Karbolfuchsin, rein

Lieferant: Thermo Fisher Scientific
Gefahr

195790050 195790250 195791000
ACRO195790050EA 43.6 EUR
ACRO195790050 ACRO195790250 ACRO195791000
Karbolfuchsin, rein
Karbolfuchsin
Formel: C₂₁H₂₂ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00143923
CAS-Nummer: 4197-24-4
EINECS: 224-086-6
UN: 2923
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
UV Lambda max (in water) about 542 nm
Suitability (for microscopy) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR