Kaliumthiocyanat 99+% zur Analyse
Lieferant: Thermo Fisher Scientific
|
Achtung
|
196585000
196580025
196580100
196582500
ACRO196585000EA
68.2
EUR
ACRO196585000
ACRO196580025
ACRO196580100
ACRO196582500
Kaliumthiocyanat 99+% zur Analyse
Kaliumthiocyanat
Formel:
KSCN Siedepunkt: 500 °C (1013 hPa) Schmelzpunkt: 170…179 °C Dichte: 1,886 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011413 CAS-Nummer: 333-20-0 EINECS: 206-370-1 Merck Index: 12,07866 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Titration Argentometric | ≥99.0 % |
pH | 5.3 to 8.7 (5 % at 20°C) |
Chloride (Cl) | ≤50 ppm |
Sulfate (SO4) | ≤50 ppm |
Ammonium (NH4) | ≤10 ppm |
Iron (Fe) | ≤2 ppm |
Sodium (Na) | ≤200 ppm |
Heavy metals (as Pb) | ≤2 ppm |
Iodine consuming subst. | ≤30 mg I2/100 g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...