Vanadium(III)chlorid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

197000250 197001000 197000050
ACRO197000250EA 116 EUR
ACRO197000250 ACRO197001000 ACRO197000050
Vanadium(III)chlorid 97%
Vanadium(III)chlorid
Formel: Cl₃V
Dichte: 3 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011454
CAS-Nummer: 7718-98-1
EINECS: 231-744-6
UN: 2475
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Dark blue to violet to black Powder
Titration Argentometric ≥96.0 %
Insoluble matter ≤0.5 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR