Natriumperchlorat 99+% zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

197120010 197122500 197120051
ACRO197120010EA 489 EUR
ACRO197120010 ACRO197122500 ACRO197120051
Natriumperchlorat 99+% zur Analyse
Sodium perchlorate
Formel: ClNaO₄
Schmelzpunkt: 482 °C
Dichte: 2,4989 to 2.4999 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003480
CAS-Nummer: 7601-89-0
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 13,08726

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Assay ≥99.0 % (on dry substance)
Loss on drying ≤2.0 % (130°C, 2 h)
pH 6.0 to 8.0 (5 % in water)
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤20 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤500 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR