Mangan(II)bromid Tetrahydrat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Mangandibromid Tetrahydrat
197130500
ACRO197130500EA
51.4
EUR
ACRO197130500
Mangan(II)bromid Tetrahydrat 98%
Mangan(II)bromid Tetrahydrat
Formel:
Br₂Mn·4H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149791 CAS-Nummer: 10031-20-6 EINECS: 236-591-9 |
Spezifikation Testergebnisse
Appearance | Pink Crystalline powder or crystals |
Titration Argentometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...